This product is now discontinued, View Related Products
Cole-Parmer Handheld Homogenizers
Mix, emulsify, shred, or chop 1- to 1000-mL biological samples
  • Dual speed motor provides versatility
These homogenizers are lightweight and simple to use—just hold homogenizer in a sample vessel, then press and hold the two-speed switch (7000 or 10,000 rpm). One-handed operation is possible with any suitable beaker or vessel having a 2 inch opening. Material is rapidly dispersed and disintegrated by the ultrahigh-frequency mechanical treatment. No sample transfer loss to transfers from a blender vessel.
Clean between samples by briefly running the rotor stator in a beaker of clean water or mild detergent.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Sample Size (mL) Min Sample Size (mL) Power (VAC)
Availability Pricing
COMPARE
200 1 110
Call For Price
COMPARE
200 1 230
Call For Price
COMPARE
1000 100 110
Call For Price
COMPARE
1000 100 230