Cole-Parmer® High-Efficiency Absolute-Rated Filter Bags, Polypropylene
Wide range of options for temperature, flow rate, and filtration quality
  • Constructed of polypropylene (PP)
  • FDA-compliant
Designed for 98% removal efficiency at the rated pore size, these bags offer three to four times the dirt-holding capacity of standard bags. In addition, they are FDA-compliant with respect to all regulations governing food-grade materials. These polypropylene absolute-rated filter bags are rated for a temperature of 200°F (93°C) maximum.
$30.75 - $66.75USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Filter Bag Size Number Flow Rate Pore Size (µm)
Availability Pricing
COMPARE
110 GPM 1
$55.25USD / Each
COMPARE
110 GPM 25
$57.50USD / Each
COMPARE
2 150 GPM 50
$30.75USD / Each
COMPARE
2 150 GPM 100
$66.75USD / Each