Cole-Parmer® High pH Double-Junction Electrodes with Integrated Assemblies
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
These electrodes are ideal for applications involving high-sodium concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Use for measurements from 0 to 14 pH. Electrodes withstand temperatures from 23 to 230ºF (-5 to 110ºC).
$261.00 - $324.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Removable High pH Double-Junction pH Electrode
$261.00USD / Each
COMPARE
Removable High pH Double-Junction pH Electrode, 100 Ohms RTD
$324.00USD / Each