Cole-Parmer High-Speed Microcentrifuge
Capable of speeds up to 16,100 x g
  • Save bench space with 9" wide centrifuge
  • Quick deceleration for shorter down time
  • Can hold up to 12 x 1.5/2.0-mL tubes
  • Control knob and bright LCD provide clear indication of time and speed
This compact microcentrifuge offers speed and force usually found on larger models. The unit delivers shortened acceleration and deceleration time with a quiet motor. The brushless, maintenance free drive system provides a powerful, smooth operation. A separate momentary spin button permits instant spin downs, without setting or disturbing any preset parameters.
$1,350.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Tube Capacity
Availability Pricing
COMPARE
15500 115 12 x 1.5/2.0 mL
$1,350.00USD / Each
COMPARE
15500 230 12 x 1.5/2.0 mL
$1,350.00USD / Each