Cole-Parmer® Hydrochloric Acid

Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$32.00 - $384.75USD / Each

18 variations of this product are available.

See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
COMPARE
Components all ACS 4000 Hydrochloric Acid, 25% (v/v), 4 L
$201.25USD / Each
COMPARE
Components all ACS 4000 Hydrochloric Acid, 0.1N (N/10), 4 L
$104.50USD / Each
COMPARE
Components all ACS 4000 Hydrochloric Acid, 0.500N (N/2), 4 L
$104.50USD / Each
COMPARE
Components all ACS 1000 Hydrochloric Acid, 0.500N (N/2), 1 L
$53.60USD / Each
COMPARE
Components all ACS 4000 Hydrochloric Acid, 1.00N, 4 L
$108.25USD / Each
COMPARE
Components all ACS 500 Hydrochloric Acid, 1.00N, 500 mL
$32.00USD / Each
COMPARE
Components all ACS 1000 Hydrochloric Acid, 1.00N, 1 L
$53.60USD / Each
COMPARE
Components all ACS 20000 Hydrochloric Acid, 1.00N, 20 L
$254.75USD / Each
COMPARE
Components all ACS 4000 Hydrochloric Acid, 6.00N, 4 L
$188.50USD / Each
COMPARE
Components all ACS 1000 Hydrochloric Acid, 6.00N, 1 L
$78.80USD / Each
COMPARE
None 500 Hydrochloric Acid, 0.1N (N/10) in Isopropyl Alcohol, 500 mL
$72.50USD / Each
COMPARE
Technical 2500 Hydrochloric Acid, Technical Grade, 20°Baumé, 2.5 L
$89.90USD / Each
COMPARE
None 1000 Hydrochloric Acid, 0.1N (N/10), 1 L
$53.60USD / Each
COMPARE
Components all ACS 1000 Hydrochloric Acid, 25% (v/v), 1 L
$76.70USD / Each
COMPARE
ACS Reagent 1000 Hydrochloric Acid, ACS Grade, 1 L
$93.60USD / Each
COMPARE
ACS Reagent 2500 Hydrochloric Acid, ACS Grade, 2.5 L
$151.50USD / Each
COMPARE
ACS Reagent 20000 Hydrochloric Acid, ACS Grade, 20 L
$384.75USD / Each
COMPARE
ACS Reagent 500 Hydrochloric Acid, ACS Grade, 500 mL
$99.00USD / Each
Showing 1- 18 of 18

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat