Cole-Parmer® IPS Portable Industrial Bench Scales
Goes anywhere with built-in rechargeable battery
  • Long 70-hour battery life means fewer recharging cycles
  • User IDs and password protection ensure security
  • Adjustable indicator is easy to see from any angle
  • Capacity tracker prevents overload conditions
  • Preset tare and selectable sample size make tasks more efficient
  • Bidirectional RS-232 interface connects with computers and printers
  • Rugged 304 stainless steel and ABS construction withstand harsh environments
  • Sealed keypad resists damage from dirt and spills
  • 2-year warranty
Industrial scales feature weighing, parts counting, percentage, checkweighing, dynamic, and accumulation functions.
ITEM(S) ON SALE
$440.10 - $684.99USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
COMPARE
15 30 115
$440.10USD / Each
$511.75USD / Each
COMPARE
50 100 115
$489.77USD / Each
$569.50USD / Each
COMPARE
75 150 115
$586.30USD / Each
$681.75USD / Each
COMPARE
150 300 115
$684.99USD / Each
$796.50USD / Each
COMPARE
15 30 220
$440.10USD / Each
$511.75USD / Each
COMPARE
50 100 220
$489.77USD / Each
$569.50USD / Each
COMPARE
75 150 220
$586.30USD / Each
$681.75USD / Each
COMPARE
150 300 220
$684.99USD / Each
$796.50USD / Each
COMPARE
25 50 115
$587.59USD / Each
$683.25USD / Each
COMPARE
25 50 220
$587.59USD / Each
$683.25USD / Each
Showing 1- 10 of 10