Cole-Parmer Jar Mills
Rugged lab mills withstand thousands of hours of operation—great for all your milling, tumbling, and polishing needs.
  • Silent operation with internal gear transmission—no chains or belts
  • Durable welded steel case with chip- and corrosion-resistant black powder coating
These jar mills are ideal for grinding laboratory or high-purity samples. High-quality, powder-coated finish protects the exterior from oils, solvents, and chemicals. Mills provide quiet, smooth operation with the 1/12-hp, direct-drive DC motor. Easy to clean and maintain, these lab mills require no tension adjustment or lubrication. Rollers and brackets are easily repositioned and secured by convenient allen-head bolts. Rollers measure 12-5/8" (32.1 cm) in length and accept jars from 2" (5.1 cm) to 10" (25.4 cm) in diameter. Maximum capacity of 30 lb (13 kg).
Low power consumption during continuous operation. Control speed from 10 to 260 rpm with front control knob, a potentiometer that adjusts power output voltage.
$4,935.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
115 / 230
$4,935.00USD / Each