Cole-Parmer Laboratory Benchtop Turbidity Meters
Front-panel dials simplify your turbidity measurements
  • Additional sample cells within easy reach—stored directly on the side of the meter
Use these meters to verify water cleanliness in public water systems and to control chemicals through water treatment. Other applications include chemically analyzing chlorides and other elements through controlled precipitation, checking filter efficiency, and isolating filter backwash problems. Meters measure particulate content of water in three ranges. All results are displayed in NTU on the 1/2"H (1.3 cm) LCD.
$1,542.00 - $1,552.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Min Turbidity Turbidity Resolution Power (VAC)
Availability Pricing
COMPARE
0 to 199.9 0.1 NTU 110
$1,542.00USD / Each
COMPARE
0 to 199.9 0.1 NTU 230
$1,552.50USD / Each