Cole-Parmer® MP-200 Series Stuart Digital Melting Point Apparatus
No more reading glass thermometers
  • Select, measure, and display the temperature digitally, ensuring accuracy and repeatability
  • Easily view the melt via magnifying lens with built-in illumination
  • Be more productive—test two samples simultaneously
  • Save time by setting a plateau temperature for initial warm-up
Designed for maximum safety and ease of use. Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test two samples simultaneously, viewing the melt via a magnifying lens with built-in illumination. Extend the back feet for the optimal viewing angle.
Set a plateau temperature for initial warm-up—unattended warm-up time frees up the user until the plateau temperature is reached. After warm-up, heating rate continues slowly reaching the melt temperature for greater accuracy.
$1,006.00 - $1,470.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Power (VAC) Temperature Resolution
Availability Pricing
COMPARE
20°C per minute; 2°C per minute to melt 120 1°C
$1,195.00USD / Each
COMPARE
20°C per minute; 2°C per minute to melt 240 1°C
$1,006.00USD / Each
COMPARE
20°C / minute to plateau; 1 and 10°C/min to melt 120 0.1°C
$1,470.00USD / Each
COMPARE
20°C / minute to plateau; 1 and 10°C/min to melt 230 0.1°C
$1,382.00USD / Each