Cole-Parmer® MS-3400 Centrifuges
A true workhorse centrifuge with versatile rotor options
  • Tube adapter options accommodate tube sizes from 0.5 to 50 mL
  • Digital keypad interface is glove-friendly
  • Quiet running centrifuge provides a pleasant work setting
This may be the only centrifuge you need—standard four-place swinging buckets
accommodate 50-mL tubes and additional rotor and adapter options accommodate
almost any tube size for your runs.
Two-line digital display shows status, time, rpm and RCF(g), and the 60-minute timer
signals when run is complete. Previous settings can be easily repeated for quick
testing. Stainless steel interior chamber is easily accessible for cleaning and
will withstand years of use.
$3,180.00 - $4,153.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Tube Capacity
Availability Pricing
COMPARE
120 4 x 50 mL
$3,180.00USD / Each
COMPARE
220 4 x 50 mL
$3,299.00USD / Each
COMPARE
120 4 x 50 mL-4 x 5 to 15 mL-4 x 3.5 to 6.5 mL-4 x 0.5 to 3 mL
$3,991.50USD / Each
COMPARE
220 4 x 50 mL-4 x 5 to 15 mL-4 x 3.5 to 6.5 mL-4 x 0.5 to 3 mL
$4,153.50USD / Each