This product is now discontinued, View Related Products
Cole-Parmer Multichannel Pipettors
Excellent accuracy and performance at an economical price
  • Fully autoclavable
  • Easy-click digital volume setting
Ergonomic, lightweight design with soft plunger movement minimizes hand fatigue. Pipettors feature streamlined, low-pressure tip ejector for easy access to narrow containers. They facilitate last drop dispensing and reverse pipetting technique with two-stage plunger operation for blow-out. Easy to calibrate and disassemble for routine servicing or autoclaving.
Adjustable-volume and multichannel pipettors feature quick, easy click-stop, digital volume settings. Individually calibrated in accordance with DIN 12650 Standards and supplied with a computer-generated calibration document supplied by the manufacturer and calibration tool.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Volume (mL) Min Volume (mL) Number Of Channels
Availability Pricing
COMPARE
0.01 0.0005 8
Call For Price
COMPARE
0.02 0.0002 8
Call For Price
COMPARE
0.05 0.0005 8
Call For Price
COMPARE
0.1 0.01 8
Call For Price
COMPARE
0.2 0.02 8
Call For Price
COMPARE
0.3 0.04 8
Call For Price
COMPARE
0.01 0.005 12
Call For Price
COMPARE
0.02 0.002 12
Call For Price
COMPARE
0.05 0.005 12
Call For Price
COMPARE
0.1 0.01 12
Call For Price
COMPARE
0.01 0.0001 12
Call For Price
COMPARE
0.3 0.04 12
Call For Price
Showing 1- 12 of 12