Cole-Parmer Normally Open SPST Switches
Install switch at pipe elbow effortlessly
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 SS, and epoxy
  • Effortlessly install with 9/16-18 UNF connections
  • Durable switch with 225°F (107°C) max temperature and 250 psi at 70°F max pressure
No need to cut a straight pipe to install your flow switch. These switches with 90° ports easily fit where a pipe elbow would be installed. Use a 50 μm filter to protect your switch. Switches include 24" (61.0 cm) L leads.
$103.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Normally open SPST flow switch; 0.1 GPM (9/16"-18 UNF)
$103.00USD / Each
COMPARE
Normally open SPST flow switch; 0.25 GPM
$103.00USD / Each
COMPARE
Normally open SPST flow switch; 0.5 GPM (9/16"-18 UNF)
$103.00USD / Each
COMPARE
Normally open SPST flow switch; 0.75 GPM
$103.00USD / Each
COMPARE
Normally open SPST flow switch; 1.0 GPM (9/16"-18 UNF)
$103.00USD / Each
COMPARE
Normally open SPST flow switch; 1.5 GPM (9/16"-18 UNF)
$103.00USD / Each