Cole-Parmer Personal Microcentrifuge Replacement Rotors
Compatible for use with the Cole-Parmer Personal Microcentrifuges
  • Rotors can quickly and easily be interchanged
These replacement rotors are compatible with the 17310-XX series Cole-Parmer personal centrifuges. The model 17310-40 rotor will hold 6 x 1.5 to 2.0 mL tubes. Model 17310-45 rotor will hold 0.4 mL strip PCR tubes. Centrifuges are sold separately.
$53.00 - $66.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Tubes Accepted Tube Sizes Accepted Description
Availability Pricing
COMPARE
6 1.5 or 2.0 mL microtubes Replacement Rotor for Personal Microcentrifuge, 6 X 1.5 mL
$66.00USD / Each
COMPARE
2 Strip tube Replacement Rotor for Personal Microcentrifuge, strip tube
$53.00USD / Each