Cole-Parmer® Polycarbonate Barbed Luer Adapters
Cole-Parmer offers a variety of barbed luer adapters to meet your application's needs. Choose from several different barb styles:
* Standard hose barb for use with flexible tubing
* Smooth bore hose barb to reduce flow turbulence and risk of line contamination
* Wide-bore hose barb for enhanced fluid path flow
* Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Both straight and elbow connectors are available. Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
$16.50 - $28.50USD / Pkg of 25
41 variations of this product are available.
See All Product Options Below
41 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Description
Availability Pricing
COMPARE
1/16 Female luer Female luer x 1/16" hose barb adapter, PC, 25/pk
$19.50USD / Pkg of 25
COMPARE
15/16 Female luer Female luer x 3/32" hose barb adapter, PC, 25/pk
$19.50USD / Pkg of 25
COMPARE
15/16 Female luer Female Luer to 3/32" wide-bore hose barb, PC, 25 per pack
$18.00USD / Pkg of 25
COMPARE
1/8 Female luer Female Luer to 1/8" wide-bore hose barb, PC, 25 per pack
$18.00USD / Pkg of 25
COMPARE
3/16 Female luer Female luer x 3/16" hose barb adapter, PC, 25/pk
$22.00USD / Pkg of 25
COMPARE
1/16 Female luer Female luer to 0.07" low-profile semi-rigid tubing barb, PC, 25 per pack
$19.50USD / Pkg of 25
COMPARE
15/16 Female luer Female luer bulkhead, 1/4-28 UNF thread, to 3/32" hose barb, PC, 25/pk
$28.50USD / Pkg of 25
COMPARE
5/8 Female luer Female luer x 1/16" hose barb, smooth bore, PC, 25/pk
$18.25USD / Pkg of 25
COMPARE
1/8 Female luer Female luer x 1/8" hose barb, smooth bore, PC, 25/pk
$18.25USD / Pkg of 25
COMPARE
15/16 Male luer lock Male luer with lock ring x 3/32" hose barb, PC, 25/pk
$21.75USD / Pkg of 25
COMPARE
5/8 Male luer lock Luer Adapters, Male Luer Lock x 1/16" ID low profile barb, PC, 25/Pk
$21.75USD / Pkg of 25
COMPARE
3/16 Male luer lock Male luer with lock ring x 3/16" hose barb, PC, 25/pk
$23.50USD / Pkg of 25
COMPARE
1/8 Male luer lock Luer Adapters, Male Luer Lock to 1/8" ID low profile barb, PC, 25/Pk
$21.75USD / Pkg of 25
COMPARE
5/8 Male luer slip Luer Adapters, Male Slip Luer to 1/16" ID, PC, 25/Pk
$19.50USD / Pkg of 25
COMPARE
15/16 Male luer slip Luer Adapters, Male Slip Luer to 3/32" ID, PC, 25/Pk
$19.50USD / Pkg of 25
COMPARE
1/8 Male luer slip Luer Adapters, Male Slip Luer to 1/8" ID, PC, 25/Pk
$23.50USD / Pkg of 25
COMPARE
15/16 Male luer slip Male luer x 3/32" hose barb (for separate lock ring), PC, 25/pk
$18.25USD / Pkg of 25
COMPARE
1/4 Male luer slip Male luer x 1/4" hose barb (for separate lock ring), PC, 25/pk
$23.50USD / Pkg of 25
COMPARE
3/16 Male luer slip Male luer slip x 3/16" hose barb, PC, 25/pk
$21.75USD / Pkg of 25
COMPARE
1/4 Male luer slip Male luer slip x 1/4" hose barb, PC, 25/pk
$21.50USD / Pkg of 25
Showing 1 - 20 of 41
Need Help with this product?
Call Us At1-847-549-7600
Live chat