Cole-Parmer® Polypropylene Microcentrifuge Tubes
Cap locking options for every application
  • Microcentrifuge tubes with graduations and frosted panels for easy labeling
  • Microbiological cleanroom controlled production (ISO Class 8)
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
These Cole-Parmer® polypropylene (PP) microcentrifuge tubes come in three cap lock options for a wide range of applications.
The double-locking caps are ideal when working with valuable samples; they prevent accidental opening during inoculation or storage and are extremely leaktight.
The click-close tubes close firmly and are ideal for heating applications.
The easy-open caps eliminate stress and sore thumbs when opening, and are best for general-purpose use and storage.
$51.50 - $80.50USD / Pkg of 500
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Style Tube Capacity (mL) Tube Color
Availability Pricing
COMPARE
Double-lock cap 0.5 Clear
$76.50USD / Pkg of 500
COMPARE
Double-lock cap 1.5 Clear
$80.50USD / Pkg of 1000
COMPARE
Double-lock cap 2 Clear
$76.50USD / Pkg of 500
COMPARE
Click Close 0.5 Clear
$63.50USD / Pkg of 500
COMPARE
Click Close 0.5 Black
$68.00USD / Pkg of 500
COMPARE
Click Close 1.5 Clear
$70.50USD / Pkg of 1000
COMPARE
Click Close 1.5 Green
$69.50USD / Pkg of 1000
COMPARE
Click Close 1.5 Blue
$69.50USD / Pkg of 1000
COMPARE
Click Close 1.5 Black
$80.50USD / Pkg of 1000
COMPARE
Click Close 2 Pink
$56.50USD / Pkg of 500
COMPARE
Click Close 2 Green
$55.50USD / Pkg of 500
COMPARE
Click Close 2 Blue
$55.50USD / Pkg of 500
COMPARE
Click Close 2 Black
$56.50USD / Pkg of 500
COMPARE
Easy Open 0.5 Clear
$51.50USD / Pkg of 500
COMPARE
Easy Open 0.5 Pink
$51.50USD / Pkg of 500
COMPARE
Easy Open 0.5 Yellow
$51.50USD / Pkg of 500
COMPARE
Easy Open 0.5 Blue
$51.50USD / Pkg of 500
COMPARE
Easy Open 1.5 Clear
$57.50USD / Pkg of 1000
COMPARE
Easy Open 1.5 Pink
$63.50USD / Pkg of 1000
COMPARE
Easy Open 1.5 Green
$63.50USD / Pkg of 1000
Showing 1 - 20 of 22