This product is now discontinued, View Related Products
Cole-Parmer Polystat High-Capacity Recirculating Chillers
Affordable – with a wide range of temperature control and cooling capacities
  • High and low temperature alarms
  • Front-mount pressure gauge for easy view of pressure readings
  • Large fill port for easy spill-proof filling
Polystat chillers are both an economical and ecological solution by allowing you to eliminate tap water waste and providing reliable liquid cooling performance for all your application requirements. Robust pump performance for closed loop recirculation up to 2.0 kW. Three pump options handle a variety of application requirements. These chillers have an intuitive digital controller with a seamless moisture-resistant keypad. Illuminated icons simplify monitoring and adjusting operation parameters. You can adjust display resolution of 1° or 0.1°C.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
700 115 7.6
Call For Price
COMPARE
600 230 7.6
Call For Price
COMPARE
1000 115 7.6
Call For Price
COMPARE
900 230 7.6
Call For Price
COMPARE
1000 115 7.6
Call For Price
COMPARE
900 230 7.6
Call For Price
COMPARE
1000 115 7.6
Call For Price
COMPARE
900 230 7.6
Call For Price
COMPARE
2000 208 to 230 18.9
Call For Price
COMPARE
1900 230 18.9
Call For Price
COMPARE
2000 208 to 230 18.9
Call For Price
COMPARE
1900 230 18.9
Call For Price
Showing 1- 12 of 12