Cole-Parmer Polystat Low-Temperature Immersion Probe Chillers
Reduce the expense of using dry ice or liquid nitrogen – excellent for rapidly cooling small volumes of liquids, Dewar-type applications
Quickly cool the contents of your bath with these immersion chillers. Digital display of bath temperature with 0.1°C resolution, and a 100Ω platinum RTD sensor for increased temperature stability. RTD temperature sensor includes a 6"L x 1/8" diameter stainless steel sheath and a 5-ft (1.5-m) cable.
Probe can be inserted into your bath at ambient temperature—there’s no need to wait until your bath reaches subzero temperatures. A generous 60" of insulated tubing (11/2" OD) is between the probe and the cabinet.
$5,350.00 - $5,770.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
-100 115 low-temperature immersion chiller; cold-finger; 120V
$5,350.00USD / Each
COMPARE
-100 230 low-temperature immersion chiller; cold-finger; 240V
$5,770.00USD / Each
COMPARE
-100 115 low-temperature immersion chiller; flex probe; 120V
$5,745.00USD / Each
COMPARE
-100 230 low-temperature immersion chiller; flex probe; 240V
$5,740.00USD / Each
COMPARE
-100 115 low-temperature immersion chiller; coil probe; 120V
$5,360.00USD / Each
COMPARE
-100 230 low-temperature immersion chiller; coil probe; 240V
$5,770.00USD / Each