Cole-Parmer Polystat Refrigerated Chillers with Immersion Probe

Quickly chill liquids in beakers, flasks or open baths
  • An economical alternative to dry ice or liquid nitrogen
  • Excellent for trapping applications, freeze drying, and rapidly cooling small volumes of liquids
All models are designed to run at maximum cooling rate.
$1,900.00 - $2,375.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
-60 115 Chiller w/ Immersion Probe, -60 to -20 C, 115V/60Hz
$1,900.00USD / Each
COMPARE
-60 230 Chiller w/ Immersion Probe, -60 to -20 C, 240V/50Hz
$1,900.00USD / Each
COMPARE
-35 115 Chiller w/ Immersion Probe, -35 to 40 C, 115V/60Hz
$2,375.00USD / Each
COMPARE
-35 230 Chiller w/ Immersion Probe, -35 to 40 C, 240V/50Hz
$2,375.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat