Cole-Parmer® IC-400 Series Standard Immersion Circulator
Turn any tank into a circulating bath featuring a large, easy-to-read digital display
  • Ensure precision due to an integral heating coil and re-circulating pump
  • Simple intuitive operation
  • Easily adjust the flow with a convenient slide control
  • For use with reservoirs up to 28 L
Smartly suited for lab and industrial use, this immersion circulator has a large backlit digital display, yet a slim overall profile requiring minimal reservoir space. From across the room, users can readily see the process value and set value, as well as heating and circulating icons. Four buttons enable easy setting of on/off/standby, point value, °F/°C readings and offset calibration value; min/max temperatures can also be easily selected. A protective housing prevents direct contact with the integral pump and heater coil yet is easy to clean and inspect. The adjustable mounting clamp attaches securely to flat and rounded tank walls.
$1,390.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
135 120 28
$1,390.00USD / Each
COMPARE
135 240 28
$1,390.00USD / Each