Cole-Parmer® Polystat® Standard Stainless Steel Open-Top Bath
Safely heat liquids with an intelligent display, simple controls and temperature protection
  • Backlit display prominently shows process and set values
  • Heating and circulating icons enhance user monitoring
  • On-screen prompts simplify functionality
Add efficiency to any environment with an efficient stainless steel bath featuring a recirculating pump, integral heating coil, drain valve, bridge and lid. The standard controller features large, universal icons and a digital display, allowing you to quickly select On/Off/Standby, enter the set-point value, choose °C or °F readings and set the offset calibration value.
Safety is enhanced by a protective housing preventing direct contact with the pump and coil, which can be easily cleaned and inspected. Rear ports can be used for external process temperature control, while Min/Max temperature is controlled by low/high limit settings.
$1,730.00 - $1,775.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
135 120 6
$1,730.00USD / Each
COMPARE
135 240 6
$1,730.00USD / Each
COMPARE
135 120 10
$1,775.00USD / Each
COMPARE
135 240 10
$1,775.00USD / Each