Cole-Parmer Precleaned EPA Amber Narrow-Mouth Glass Jugs
Certified clean for worry-free sampling
  • Include certificate of conformance to EPA specifications
  • Sample analysis labels included (unattached)
Glass jugs are processed and certified to meet the requirements of the current US EPA “Specifications and Guidance for Contaminant-Free Sample Containers” document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate and nitrite analyses. Include PTFE-lined caps.
$52.50 - $60.00USD / Case of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material
Availability Pricing
COMPARE
2.5 Glass
$60.00USD / Case of 6
COMPARE
4 Glass
$52.50USD / Case of 4
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat