Cole-Parmer Precleaned EPA Amber Wide-Mouth Glass Bottles
Certified clean for worry-free sampling
  • Include certificate of conformance to EPA specifications
  • Sample analysis labels included (unattached)
Glass bottles are processed and certified to meet the requirements of the current US EPA “Specifications and Guidance for Contaminant-Free Sample Containers” document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate and nitrite analyses. Include PTFE-lined caps.
$41.00 - $82.00USD / Case of 12
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 38-400 Glass
$41.00USD / Case of 12
COMPARE
250 45-400 Glass
$45.00USD / Case of 12
COMPARE
500 53-400 Glass
$63.50USD / Case of 12
COMPARE
1000 53-400 Glass
$82.00USD / Case of 12
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat