Cole-Parmer Precleaned EPA Clear Wide-Mouth Glass Septa Jars
Certified clean for worry-free sampling
  • Jars include certificate of conformance to EPA specifications
  • Includes PTFE-coated silicone septas and caps
Glass Septa Jars are processed and certified to meet the requirements of the current US EPA “Specifications and Guidance for Contaminant-Free Sample Containers” document for purgeable volatile organic analysis. Include PTFE-lined silicone septa bonded to open-top caps.
$99.00 - $103.00USD / Case of 24
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material
Availability Pricing
COMPARE
60 Glass
$99.00USD / Case of 24
COMPARE
125 Glass
$103.00USD / Case of 24
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat