Cole-parmer® Premium Luer Fittings
Full material traceability to ensure process integrity
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of options including luer, thread, and hose barb.
$14.50 - $66.00USD / Pkg of 25
60 variations of this product are available.
See All Product Options Below
60 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Connection Type-Port 2
Availability Pricing
COMPARE
3/32 Female luer Hose barb
$16.75USD / Pkg of 25
COMPARE
15/16 Female luer Hose barb
$18.25USD / Pkg of 25
COMPARE
5/32 Female luer Hose barb
$22.50USD / Pkg of 25
COMPARE
3/16 Female luer Hose barb
$21.75USD / Pkg of 25
COMPARE
1/16 Female luer Hose barb
$29.50USD / Pkg of 25
COMPARE
1/8 Female luer Hose barb
$28.50USD / Pkg of 25
COMPARE
15/16 Female luer Hose barb
$20.00USD / Pkg of 25
COMPARE
Female luer Female luer
$37.00USD / Pkg of 25
COMPARE
Female luer 10-32 UNF thread
$40.00USD / Pkg of 25
COMPARE
Female luer 10-32 special tapered thread
$40.00USD / Pkg of 25
COMPARE
Female luer Female luer
$55.00USD / Pkg of 25
COMPARE
1/16 Male luer lock Hose Barb
$16.75USD / Pkg of 25
COMPARE
1/8 Male luer lock Hose barb
$21.75USD / Pkg of 25
COMPARE
11/64 Male luer lock Hose barb
$23.50USD / Pkg of 25
COMPARE
15/16 Male luer slip Hose barb
$20.00USD / Pkg of 25
COMPARE
1/8 Male luer slip Hose barb
$18.25USD / Pkg of 25
COMPARE
1/8 Male luer slip Hose barb
$18.25USD / Pkg of 25
COMPARE
3/16 Male luer slip Hose barb
$20.00USD / Pkg of 25
COMPARE
Male luer lock 10-32 special tapered thread
$29.50USD / Pkg of 25
COMPARE
Male luer lock 1/8-27 NPT thread
$51.00USD / Pkg of 25
Showing 1 - 20 of 60
Need Help with this product?
Call Us At1-847-549-7600
Live chat