Cole-Parmer® Psychrometers
A completely portable instrument which indicates wet and dry bulb temperatures for relative humidity and dew point measurements
  • Greater than 1% accuracy for RH determination
  • Greater than 1/4% accuracy on thermometers
  • 1000 readings on 3 «D» cells
  • Variable intensity and built-in illumination
  • Requires no whirling or special technique
  • Meets federal specification GG-P-725B, Type III, Style B
Psychrometers measure relative humidity without agitation. To use, turn on the fan and let it run for 1-1/2 to 2 minutes. Then, subtract wet bulb reading from dry bulb reading and use the included slide rule and tables to calculate the RH. The fan is powered by three size D batteries (order separately). This operation, when compared to the conventional sling psychrometers, affords a safer, convenient, and more accurate method of RH determination.
$292.00 - $295.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Battery-Operated Psychrometer, Farenheit Scale, 0 to 120°F, 10 to 100% RH
$292.00USD / Each
COMPARE
Battery-Operated Psychrometer, Celsius Scale, -15 to 45°C, 10 to 100% RH
$295.25USD / Each