This product is now discontinued, View Related Products

Cole-Parmer rotary vane pumps

Wide range of free-air capacities to match specific applications
  • Wide range of free-air capacities to match specific applications
  • Portable direct-driven high vacuum pumps—ideal for freeze dryers, vacuum ovens, and concentrators
  • Convenient inlet barb connection accepts two sets of tubing ID.
  • Gas ballast valve inhibits condensation of vapors—close for high-vacuum applications, open for applications
  • with moist vapors.
  • Isolation valve activates on power loss to prevent oil suckback and isolate vacuum chamber.
  • Time specific warning tag alerts you to change oil.
What’s included: user activated tag to encourage appropriate oil change interval, 3 — ft rubber vacuum hose, ISO — to — hose adapter kit, and 40 mL of premium vacuum pump oil.
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Free Air Capacity (CFM) Max Vacuum (torr) Power (VAC)
Availability Pricing
COMPARE
1.8 2. x 10-3 115
Call For Price
COMPARE
1.5 2. x 10-3 230
Call For Price
COMPARE
2.6 1 x 10-4 115 / 230
Call For Price
COMPARE
2.6 1 x 10-4 115
Call For Price
COMPARE
3.8 1 x 10-4 115 / 230
Call For Price
COMPARE
3.8 1 x 10-4 115
Call For Price
COMPARE
6.1 1 x 10-4 115 / 230
Call For Price
COMPARE
6.1 1 x 10-4 115
Call For Price
COMPARE
7.7 3 x 10-4 115
Call For Price
COMPARE
6.4 3 x 10-4 230
Call For Price
COMPARE
11.3 5 x 10-4 230
Call For Price
COMPARE
9.3 5 x 10-4 230
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat