EW-98965-11 Mfr # V12Z10
COMPARE Silicone Rotational Viscosity Standard Oil, 5 mPa*s; 600 mL/bottle
9896511 EW-98965-12 Mfr # V12Z14
COMPARE Silicone Rotational Viscosity Standard Oil, 10 mPa*s; 600 mL/bottle
9896512 EW-98965-13 Mfr # V12Z18
COMPARE Silicone Rotational Viscosity Standard Oil, 50 mPa*s; 600 mL/bottle
9896513 EW-98965-14 Mfr # V12Z20
COMPARE Silicone Rotational Viscosity Standard Oil, 75 mPa*s; 600 mL/bottle
9896514 EW-98965-16 Mfr # V12Z22
COMPARE Silicone Rotational Viscosity Standard Oil, 100 mPa*s; 600 mL/bottle
9896516 EW-98965-17 Mfr # V12Z23
COMPARE Silicone Rotational Viscosity Standard Oil, 250 mPa*s; 600 mL/bottle
9896517 EW-98965-18 Mfr # V12Z24
COMPARE Silicone Rotational Viscosity Standard Oil, 300 mPa*s; 600 mL/bottle
9896518 EW-98965-19 Mfr # V12Z26
COMPARE Silicone Rotational Viscosity Standard Oil, 500 mPa*s; 600 mL/bottle
9896519 EW-98965-21 Mfr # V12Z30
COMPARE Silicone Rotational Viscosity Standard Oil, 1000 mPa*s; 600 mL/bottle
9896521 EW-98965-22 Mfr # V12Z34
COMPARE Silicone Rotational Viscosity Standard Oil, 5000 mPa*s; 600 mL/bottle
9896522 EW-98965-23 Mfr # V12Z36
COMPARE Silicone Rotational Viscosity Standard Oil, 10000 mPa*s; 600 mL/bottle
9896523 EW-98965-24 Mfr # V12Z38
COMPARE Silicone Rotational Viscosity Standard Oil, 12500 mPa*s; 600 mL/bottle
9896524 EW-98965-26 Mfr # V12Z42
COMPARE Silicone Rotational Viscosity Standard Oil, 30000 mPa*s; 600 mL/bottle
9896526 EW-98965-27 Mfr # V12Z46
COMPARE Silicone Rotational Viscosity Standard Oil, 60000 mPa*s; 600 mL/bottle
9896527 EW-98965-28 Mfr # V12Z50
COMPARE Silicone Rotational Viscosity Standard Oil, 100000 mPa*s; 600 mL/bottle
9896528 EW-98965-52 Mfr # V12010N
COMPARE Small Sample Adapter with Circulation Jacket
9896552 COMPARE Small Sample Adapter (Accessories for Rotational Viscometers)
9896553 EW-98965-54 Mfr # V12013N
COMPARE Low Viscosity Adapter with Circulation Jacket
9896554 COMPARE Rotational Viscometer Low Viscosity Adapter
9896555 COMPARE Rotational Viscometer Helix Path Drive Unit, HELDAL
9896556