This product is now discontinued, View Related Products
Cole-Parmer Shop Microscopes 3077
Take anywhere on the job - ideal for lab, field, or shop work
Small size and rugged carrying case make it easy to take these shop microscopes anywhere! The cast aluminum frame with gray enamel finish measures only 9''H. Wide-field 10x eyepiece features a direct-reading reticle with both inch and metric scales. Simply turn the eyepiece to bring the reticle into sharp focus - no calibration is needed. Dual knobs with rack and pinion focusing let you quickly refine the image. Penlight illuminator is fully adjustable in all directions by means of a ball joint; remove it for use elsewhere.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Maximum Magnification
Availability Pricing
COMPARE
20x
Call For Price
COMPARE
40x
Call For Price
COMPARE
60x
Call For Price
COMPARE
100x