Cole-Parmer Single-Head Chemical Feed Diaphargm Pumps
  • Easily prime over long distances
  • Low strokes/minute rate increases diaphragm life
  • Overload-protected, fan-cooled, shaded-pole gear motor (1/35 hp) is permanently lubricated
  • Sealed VALOX® housing resists harsh weather conditions
Pumps come complete with foot valve/strainer, polypropylene antisiphon valve, vinyl inlet and polyethylene outlet tubing, mounting hardware, and 6-ft, three-wire cord and plug.
$578.00 - $603.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Flow Rate (Strokes/min) Max Pressure (PSI)
Availability Pricing
COMPARE
22 14 125
$603.00USD / Each
COMPARE
46 30 125
$578.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat