Oakton® by Cole-Parmer® Special-Purpose Electrodes
Withstand high temperatures and chemical attack
  • Exhibit minimal solution carryover
Glass-body combination electrodes feature refillable references, except for 05998-30 which features a high-temperature KNO3 gel. All have BNC connectors.
ITEM(S) ON SALE
$158.40 - $225.90USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH electrode, combination, spear tip, sealed, glass body, BNC
$225.90USD / Each
$251.00USD / Each
COMPARE
pH electrode, combination, double-junction, KNO3 gel, glass body, BNC
$158.40USD / Each
$176.00USD / Each