Cole-Parmer Specific Gravity Precision Thermo-Hydrometers for Liquids Lighter than Water
  • NIST Traceable
  • Blue Spirit
  • Non-Mercury
  • Temperature of Standardization 60°F/60°F
  • Individually Serialized
These thermo-hydrometers feature non-mercury performance liquid and conform to the accuracy requirements for ASTM thermo-hydrometers. Each thermo-hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST. Made in the USA.
ITEM(S) ON SALE
$54.99 - $55.93USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0.700/0.810 Specific Gravity Combined Form Thermo-Hydrometer
$54.99USD / Each
$64.70USD / Each
COMPARE
0.800/0.910 Specific Gravity Combined Form Thermo-Hydrometer
$55.93USD / Each
$65.80USD / Each
COMPARE
0.900/1.000 Specific Gravity Combined Form Thermo-Hydrometer
$54.99USD / Each
$64.70USD / Each