Cole-Parmer® Spex® Grinding Dishes for SM-300 ShatterBox®
Handles allow you to easily lift your sample container in and out of the ShatterBox® Verastile Ring and Puck Mill
  • Suitable for slurry grinding
  • Dish-shaped grinding container swings in a tight high-speed circle to crush samples to analytical fineness
Typical samples include: Cements, Soils, Ceramics, Slag, Rocks, Minerals, Ores, Pharmaceuticals, Sulfur Pellets, Dried Plant Material
$2,482.00 - $12,073.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
COMPARE
270 Hardened steel Grinding Container with Handles, Hardened Steel; 270 mL
$2,482.00USD / Each
COMPARE
240 Tungsten carbide Grinding Container with Handles, Tungsten Carbide; 240 mL
$12,073.00USD / Each
COMPARE
170 Alumina ceramic Grinding Container with Handles, Alumina Ceramic; 170 mL
$3,814.00USD / Each
COMPARE
170 Zirconia Grinding Container with Handles, Zirconia; 170 mL
$7,020.00USD / Each
COMPARE
650 Hardened steel Grinding Container with Handles, Hardened Steel; 650 mL
$3,510.00USD / Each