Cole-Parmer® SSH-400 Series Sonic Shifters
Sonic sieving action combined with two-way (horizontal and vertical) tapping provide the most accurate and reliable particle separation
  • Sonic energy pulses reorient particles to the mesh surface for precise, repeatable separation of granular materials and fine powders
  • Easy-to-use control panel allows fully customizable testing profiles
  • Enclosed design helps confine dust particles
  • Set operating time up to 99.9 minutes
Sonic sifters deliver 3600 sonic pulses/min oscillation inside the sieve stack to provide precise, dependable separation of powders, granular materials, and minerals down to 5 µm. Vertical and horizontal tapping actions ensure maximum particle movement to separate hard-to-separate materials while reducing sample clumping. Easily set operating parameters using a control panel with a large LED display. Set vibration and amplitude, ramp and dwell, test time, tapping action as well as 10 fully customizable profiles to ensure accurate and repeatable test results.
$8,550.00 - $9,901.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Sieves Size Accepted (OD) Description
Availability Pricing
COMPARE
115 / 230 3\" Accepts 3\" Diameter Sieves; 115 VAC, 60 Hz or 230 VAC, 50 Hz
$8,550.00USD / Each
COMPARE
115 / 230 8\" Accepts 8\" Diameter Sieve; 115 VAC, 60 Hz or 230 VAC, 50 Hz
$9,901.00USD / Each