This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heater, Combination Heating Blocks
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Combination blocks are single size blocks, are available in 3 different combinations, and each hold 3 different size tubes.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Combination Block -Tube / Vial Size Accepted Description
Availability Pricing
COMPARE
16 12/13, 6 & 25 mm Single Block Combination, Test Tubes
Call For Price
COMPARE
9 1.5, 15 & 50 mL Single Block Combination, Centrifuge Tubes
Call For Price
COMPARE
21 0.5, 1.5 & 2 mL Single Block Combination, Micro Centrifuge Tubes