Cole-Parmer StableTemp Dry Block Heater, Stainless Steel Sand Baths Heating Blocks and Accessories
Stainless Steel Sand Baths are ideal for irregular vessels or to hold a variety of different size vessels at one time. Constructed of stainless steel for superior corrosion resistance. Designed to hold sand, stainless steel shot, or non-volatile liquids. Fits into the Cole Parmer Stable Temp Dry Block Heaters in place of a modular block. Select the appropriate capacity for the size block heater.
$86.50 - $359.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
COMPARE
2 1/2 Stainless Steel Sand Baths, 1 Block Models
$303.00USD / Each
COMPARE
3 3/4 Stainless Steel Sand Baths, 2 Block Models
$310.00USD / Each
COMPARE
3 3/4 Stainless Steel Sand Baths, 3 Block Models
$333.00USD / Each
COMPARE
5 51/64 Stainless Steel Sand Baths, 4 Block Models
$347.00USD / Each
COMPARE
5 51/64 Stainless Steel Sand Baths, 6 Block Models
$359.00USD / Each
COMPARE
Sand for Stainless Steel Bath, 1 Lb
$86.50USD / Each
COMPARE
Stainless steel shot for stainless steel bath. 1 lb
$112.00USD / Each
Showing 1- 7 of 7
Need Help with this product?
Call Us At1-847-549-7600
Live chat