Cole-Parmer StableTemp Economy Mechanical Convection Ovens
Forced-air convection with PID temperature control
  • Run simple programs — 3 preset temperature settings, delayed on/off, and timed runs
  • Safely operate in any lab with the over-temperature and door-open alarms
  • Resist corrosion—Incoloy® sheathed heating element and stainless steel interior
These economical forced-air convection ovens feature microprocessor PID control with auto-tuning and calibration functions commonly found on more expensive ovens. Digital timer up to 99 hours, 59 minutes and a delayed on/off feature offer the convenience of unattended operation. The dual-wall door opens 180° for greater access to the chamber. Rounded inner chamber corners offer easy cleaning and superior air circulation. Two 1.5" (3.8 cm) diameter vents are located on the top of the unit—adjustable ventilation provides control of inner air and vapor circulation.
$2,320.00 - $3,400.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
COMPARE
1.8 120
$2,320.00USD / Each
COMPARE
3.5 120
$2,840.00USD / Each
COMPARE
5.3 120
$3,400.00USD / Each
COMPARE
1.8 230
$2,320.00USD / Each
COMPARE
3.5 230
$2,840.00USD / Each
COMPARE
5.3 230
$3,400.00USD / Each
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat