Cole-Parmer StableTemp Hot Plates
Precise control and enhanced safety
  • Hot Surface Warning safety symbol remains lit until top plate cools below 50°C
  • Contoured display diverts liquid spills away from internal components
  • Choose corrosion-resistant ceramic for high-temperature applications or rugged aluminum for temperature uniformity
  • Overtemperature protection system cuts off power to unit in overheat condition, protecting both user and sample
  • Optional splash guard (order separately) protects you and your sample from mishaps
$268.00 - $854.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Material Description
Availability Pricing
COMPARE
100 to 120 Ceramic Ceramic Top Hot Plate, 4 x 4, 100-120 VAC
$268.00USD / Each
COMPARE
230 Ceramic Ceramic Top Hot Plate, 4 x 4, 230 VAC
$341.00USD / Each
COMPARE
100 to 120 Ceramic Ceramic Top Hot Plate, 7 x 7, 100-120 VAC
$297.00USD / Each
COMPARE
230 Ceramic Ceramic Top Hot Plate, 7" x 7", 230 V
$336.00USD / Each
COMPARE
100 to 120 Aluminum Aluminum Top Hot Plate, 7 x 7, 100-120 VAC
$359.00USD / Each
COMPARE
230 Aluminum Aluminum Top Hot Plate, 7 x 7, 230 VAC
$431.00USD / Each
COMPARE
100 to 120 Ceramic Ceramic Top Hot Plate, 10 x 10, 100-120 VAC
$470.00USD / Each
COMPARE
230 Ceramic Ceramic Top Hot Plate, 10 x 10, 230 VAC
$536.00USD / Each
COMPARE
100 to 120 Ceramic Advanced Ceramic Top Hot Plate, 7 x 7, 100-120 VAC
$758.00USD / Each
COMPARE
230 Ceramic Advanced Ceramic Top Hot Plate, 7 x 7, 230 VAC
$854.00USD / Each
Showing 1- 10 of 10