Cole-Parmer Sterile PETG Square Media Bottles
Leakproof, shatterproof, optically clear, and gamma sterilized
  • PETG is a safe alternative to glass—shatterproof
  • Secure your samples with leakproof seals (included)
  • Molded-in graduations—ideal for reference filling
  • Gamma irradiated for SAL 10-6 sterility
Store buffers, culture media, and other critical fluids in these sterile bottles. Square shape saves precious shelf space; bottles can be placed closer together.
Bottles come in case quantities, and smaller packs within. Each case is specially packaged to maintain sterility—multi-barrier packaging.
$287.00 - $441.00USD / Case of 72
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Min Graduation (mL)
Availability Pricing
COMPARE
125 25 25
$441.00USD / Case of 144
COMPARE
250 25 25
$387.00USD / Case of 96
COMPARE
500 50 50
$361.00USD / Case of 72
COMPARE
1000 50 50
$287.00USD / Case of 48
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat