Cole-Parmer Sterile Vacuum Filtration Systems
Ideal for sterile filtration of cell culture media
  • Polyethersulfone (PES) membrane provides fast flow rates and low binding
  • Features a 0.22 μm PES membrane pore
  • Sterilized by gamma irradiation and packaged in individual bags
$112.00 - $113.00USD / Case of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Membrane Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
500 75 0.22
$112.00USD / Case of 12
COMPARE
50 50 0.22
$113.00USD / Case of 12
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat