Foxx Life Sciences Storage Carboys, Polypropylene
Secure grips make lifting, filling, and pouring easy
  • Easy-to-read metric graduations are certified to ±5% accuracy
  • Wide-mouth neck simplifies filling and cleaning
  • Ergonomic body design maximizes bench and shelf space
  • Large top handles offer full hand grip
  • Interchangeable leak-proof top connections
  • BPA-free, USP Class VI, FDA-grade autoclavable resin offers chemical resistance and heat stability
The easy-to-grip polypropylene storage carboys allow you to securely hold the carboy while lifting, filling, and pouring. The carboys feature easy-to-read metric graduation marks and molded-in material and volume identification markings. Some models offer an autoclavable PP spigot.
$62.60 - $667.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap Material Cap / Stopper Size
Availability Pricing
Foxx Life Sciences Carboy, Dark Amber Polypropylene, 83B Cap; 5L EW-06035-46 Mfr # 15N-1111-OEM
COMPARE
5 Polypropylene (PP) 83B
$108.25USD / Each
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap; 2.5 L EW-06059-49 Mfr # 155-0111-OEM
COMPARE
2.5 Polypropylene (PP) 80 mm
$62.60USD / Each
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap; 5 L EW-06059-50 Mfr # 155-1111-OEM
COMPARE
5 Polypropylene (PP) 80 mm
$69.10USD / Each
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap; 10 L EW-06059-52 Mfr # 155-2111-OEM
COMPARE
10 Polypropylene (PP) 80 mm
$119.75USD / Each
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap; 20 L EW-06059-55 Mfr # 155-3111-OEM
COMPARE
20 Polypropylene (PP) 80 mm
$136.00USD / Each
Foxx Life Sciences Carboy, Polypropylene, 120 mm Cap; 20 L EW-06059-56 Mfr # 155-3121-OEM
COMPARE
20 Polypropylene (PP) 120 mm
$140.75USD / Each
Foxx Life Sciences Carboy, Polypropylene, 120 mm Cap; 40 L EW-06059-60 Mfr # 155-5121-OEM
COMPARE
40 Polypropylene (PP) 120 mm
$232.25USD / Each
Foxx Life Sciences Carboy, Polypropylene, 120 mm Cap; 75 L EW-06059-63 Mfr # 155-7121-OEM
COMPARE
75 Polypropylene (PP) 120 mm
$667.00USD / Each
Showing 1- 8 of 8