Cole-Parmer® Sulfuric Acid

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$88.00 - $210.50USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
COMPARE
Components all ACS 4000 Sulfuric Acid, Babcock, 1.825 Specific Gravity; (4 L)
$200.25USD / Each
COMPARE
ACS Reagent 1000 Sulfuric Acid ACS Grade; (1 L)
$210.50USD / Each
COMPARE
ACS Reagent 2500 Sulfuric Acid ACS Grade, Glass Bottle; (2.5 L)
$187.50USD / Each
COMPARE
ACS Reagent 2500 Sulfuric Acid ACS Grade, Safety-Coated Glass Bottle; (2.5 L)
$156.50USD / Each
COMPARE
ACS Reagent 500 Sulfuric Acid ACS Grade, Glass Bottle; (500 mL)
$88.00USD / Each
COMPARE
ACS Reagent 500 Sulfuric Acid ACS Grade, Safety-Coated Glass Bottle; (500 mL)
$108.75USD / Each
COMPARE
Technical 2500 Sulfuric Acid, Technical Grade, 66 Degree Be; (2.5 L)
$140.50USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat