This product is now discontinued, View Related Products

Cole-Parmer ThinTouch™ Nitrile Gloves

Enhanced tactile sensitivity for a superior “touch & feel” experience
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

  • Advanced nitrile formulation with soft finish delivers optimal comfort and dexterity
  • Reliable protection against a wide range of chemicals
  • Textured fingertips ensure a secure grip in wet or dry conditions
  • High tensile strength for excellent tear and puncture resistance
These gloves are soft, latex-free and powder-free. Available in 9.5” length and four sizes – small, medium, large and x-large. Textured fingertips enhance wet or dry grip. Ambidextrous with beaded cuffs.
AQL: 1.5
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size Description
Availability Pricing
COMPARE
9 1/2 Small ThinTouch™ Nitrile Blue Gloves, 9.5, Small 1000/CS
Call For Price
COMPARE
9 1/2 Medium ThinTouch™ Nitrile Blue Gloves, 9.5, Medium 1000/CS
Call For Price
COMPARE
9 1/2 Large ThinTouch™ Nitrile Blue Gloves, 9.5, Large 1000/CS
Call For Price
COMPARE
9 1/2 Small ThinTouch™ Nitrile Blue Gloves, 9.5, Small 100/Box
Call For Price
COMPARE
9 1/2 Medium ThinTouch™ Nitrile Blue Gloves, 9.5, Medium 100/Box
Call For Price
COMPARE
9 1/2 Large ThinTouch™ Nitrile Blue Gloves, 9.5, Large 100/Box
Call For Price
COMPARE
9 1/2 X-Large ThinTouch™ Nitrile Blue Gloves, 9.5, X-Large 100/Box
Call For Price
COMPARE
9 1/2 X-Large ThinTouch™ Nitrile Blue Gloves, 9.5, X-Large 1000/CS
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat