Cole-Parmer® Triton X-100® Wetting Agent
Reagents and solutions to meet your lab and research needs
  • This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$35.10 - $117.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
500 Triton® X-100 Wetting Agent, 500 mL
$90.00USD / Each
COMPARE
1000 Triton® X-100 Wetting Agent, 1 L
$117.50USD / Each
COMPARE
120 Triton® X-100 Wetting Agent, 120 mL
$35.10USD / Each
COMPARE
250 Triton® X-100 Wetting Agent, 250 mL
$65.00USD / Each