This product is now discontinued, View Related Products
Cole-Parmer Uncontrolled Heating Mantles
Heat samples safely, efficiently, and faster
  • Powder-coated aluminum case is cool to the touch and easy to clean
  • Heats to maximum temperature of 842°F (450°C)
Incorporating a flexible heating element, the uncontrolled heating mantles allow for efficient heat transfer and fast boiling times of liquid samples in round-bottom flasks between 50 and 5000 mL capacity. Ceramic fiber thermal insulation concentrates the heat on the glassware and keeps the aluminum powder-coated case stable and cool. The uncontrolled mantles require the use of an external temperature controller."
Call For Price
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Max Flask Size (mL) Power (VAC)
Availability Pricing
COMPARE
50 120
Call For Price
COMPARE
50 230
Call For Price
COMPARE
100 120
Call For Price
COMPARE
100 230
Call For Price
COMPARE
250 120
Call For Price
COMPARE
250 230
Call For Price
COMPARE
500 120
Call For Price
COMPARE
500 230
Call For Price
COMPARE
1000 120
Call For Price
COMPARE
1000 230
Call For Price
COMPARE
2000 120
Call For Price
COMPARE
2000 230
Call For Price
COMPARE
5000 120
Call For Price
COMPARE
5000 230
Call For Price
Showing 1- 14 of 14