Cole-Parmer Universal Input Panel Meters
Save and recall memory for up to 10 years
  • Front panel meets NEMA 4X and IP65 standards for dust and water resistance
  • 1/8-DIN meters accept current, voltage, thermocouple, and RTD inputs
  • Large, easy-read LED display has adjustable intensity for changing light conditions
Select input/sensor type, decimal point, units of measure, scale, and alarms quickly — simple push-button programming without cryptic coding. Programming and setup can be performed with either the front-panel push buttons, the included MasterView® software, or by using the copy function. Collect only the data you need. Data can be easily imported into spreadsheets or other applications.
$279.00 - $532.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Control Output 1 DIN Size Description
Availability Pricing
COMPARE
None 1/8 Universal Input Panel Meter, 0.56\"/85-265VAC
$279.00USD / Each
COMPARE
None 1/8 Universal Input Panel Meter, 0.56\"/24VDC/85-265VAC
$325.00USD / Each
COMPARE
2 relays 1/8 Universal Input Panel Meter, 0.56\"/2-SPDT/85-265VAC
$353.00USD / Each
COMPARE
2 relays 1/8 Universal Input Panel Meter, 0.56\"/2-SPDT/24VDC/85-265VAC
$399.00USD / Each
COMPARE
4-20 mA 1/8 Universal Input Panel Meter, 0.56\"/4-20mA/85-265VAC
$388.00USD / Each
COMPARE
4-20 mA 1/8 Universal Input Panel Meter, 0.56\"/4-20mA/24VDC/85-265VAC
$410.00USD / Each
COMPARE
None 1/8 Universal Input Panel Meter, 0.56\"/12-36VDC
$337.00USD / Each
COMPARE
2 relays 1/8 Universal Input Panel Meter, 0.56\"/2-SPDT/12-36VDC
$410.00USD / Each
COMPARE
None 1/8 Universal Input Panel Meter, 1.2\"/85-265VAC
$389.00USD / Each
COMPARE
None 1/8 Universal Input Panel Meter, 1.2\"/24VDC/85-265VAC
$435.00USD / Each
COMPARE
2 relays 1/8 Universal Input Panel Meter, 1.2\"/2-SPDT/24VDC/85-265VAC
$509.00USD / Each
COMPARE
4-20 mA 1/8 Universal Input Panel Meter, 1.2\"/4-20mA/85-265VAC
$498.00USD / Each
COMPARE
4-20 mA 1/8 Universal Input Panel Meter, 1.2\"/4-20mA/24VDC/85-265VAC
$520.00USD / Each
COMPARE
None 1/8 Universal Input Panel Meter, 1.2\"/12-36VDC
$447.00USD / Each
COMPARE
2 relays 1/8 Universal Input Panel Meter, 1.2\"/2-SPDT/12-36VDC
$520.00USD / Each
COMPARE
4-20 mA 1/8 Universal Input Panel Meter, 1.2\"/4-20mA/12-36VDC
$532.00USD / Each
Showing 1- 16 of 16