Cole-Parmer® Vinyl Tubing
Applications: General-purpose lab use.
Characteristics: Fair chemical and abrasion resistance, odorless, nontoxic, and transparent.
Certification: Tubing is manufactured according to GMP.
Temperature range: –45 to 180°F
(–43 to 82°C)
Sterilize: Ethylene oxide.
$19.00 - $416.00USD / Pkg of 1
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
40 1/16 1/8
$19.00USD / Pkg of 1
COMPARE
59 1/8 1/4
$21.75USD / Pkg of 1
COMPARE
51 3/16 5/16
$24.25USD / Pkg of 1
COMPARE
46 3/16 7/16
$94.50USD / Pkg of 1
COMPARE
51 1/4 3/8
$29.00USD / Pkg of 1
COMPARE
55 1/4 7/16
$81.50USD / Pkg of 1
COMPARE
64 1/4 1/2
$83.00USD / Pkg of 1
COMPARE
46 5/16 7/16
$29.50USD / Pkg of 1
COMPARE
55 5/16 1/2
$93.00USD / Pkg of 1
COMPARE
41 3/8 1/2
$32.50USD / Pkg of 1
COMPARE
46 3/8 9/16
$107.00USD / Pkg of 1
COMPARE
55 3/8 5/8
$144.00USD / Pkg of 1
COMPARE
28 1/2 5/8
$41.50USD / Pkg of 1
COMPARE
36 1/2 11/16
$130.00USD / Pkg of 1
COMPARE
41 1/2 3/4
$87.50USD / Pkg of 1
COMPARE
32 5/8 13/16
$165.00USD / Pkg of 1
COMPARE
36 5/8 7/8
$104.00USD / Pkg of 1
COMPARE
32 3/4 1
$112.00USD / Pkg of 1
COMPARE
23 1 1 1/4
$158.00USD / Pkg of 1
COMPARE
41 1 1/4 1 3/4
$416.00USD / Pkg of 1
Showing 1- 20 of 20