Cole-Parmer® VPS-200 Series Air Admiral Vacuum Pumps
Compact size and integral handle make pump easily transportable
  • Economical vacuum pump serves as pressure or vacuum source with an excellent price to performance ratio
  • Low-friction piston runs cool and quiet for a long service life with a consistent performance
This portable vacuum pump is designed for versatility with a tough, lightweight plastic housing and integral carrying handle. The long-wearing motor delivers performance characteristics suitable for a wide range of applications.
Note: Do not pump liquids or immerse pump. Avoid organic, basic, and acidic vapors.
$495.00 - $769.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Power (VAC)
Availability Pricing
COMPARE
0.37 21.3 115
$495.00USD / Each
COMPARE
0.31 21.3 230
$575.00USD / Each
COMPARE
0.37 21.3 120
$755.00USD / Each
COMPARE
0.31 21.3 230
$769.00USD / Each