Cole-Parmer VS-3400 Variable Speed Small Benchtop Centrifuges

Ideal for sample spinning with flat separation meniscus line
  • Stainless steel guard bowl and lid liner for robust longevity
  • Real-time digital display for RPM, RCF, and run time
  • Store your settings for easy runs
These centrifuges variable speed from 1000 to 3400 rpm - ensuring a clean and complete separation of cellular components yielding a superior specimen. A unique zero rpm lid- latch safety system keeps the lid closed while the rotor is in motion. All units have real-time two-line digital display that shows status, time, and speed (rpm and RCF). Stainless steel rotor chamber is accessible for easy cleaning and disinfection.
$2,685.00 - $2,850.50USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Power (VAC) Tube Capacity
Availability Pricing
COMPARE
1825 115 6 x 2 to 15 mL
$2,850.50USD / Each
COMPARE
1825 220 6 x 2 to 15 mL
$2,850.50USD / Each
COMPARE
1740 115 24 x 2 to 10 mL-16 x 15 mL
$2,685.00USD / Each
COMPARE
1740 220 24 x 2 to 10 mL-16 x 15 mL
$2,685.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat