Colored Polyolefin Heat-Shrink Tubing
Shrink ratio: 2:1
Recovery temperature: 275degF (135degC)
Characteristics: Transparent, heavy yet flexible.
Certification: UL/CSA, VW-1, military spec MIL-I-23053/5, Class 1 & 3
$9.00 - $57.50USD / Pkg of 1
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
3/32
$12.40USD / Pkg of 1
COMPARE
1/8
$10.10USD / Pkg of 1
COMPARE
3/16
$12.90USD / Pkg of 1
COMPARE
3/8
$16.40USD / Pkg of 1
COMPARE
1/2
$19.10USD / Pkg of 1
COMPARE
1
$49.75USD / Pkg of 1
COMPARE
1/25
$11.30USD / Pkg of 1
COMPARE
1/16
$9.80USD / Pkg of 1
COMPARE
3/32
$10.50USD / Pkg of 1
COMPARE
3/16
$11.60USD / Pkg of 1
COMPARE
1/4
$14.60USD / Pkg of 1
COMPARE
3/8
$15.60USD / Pkg of 1
COMPARE
3/4
$47.75USD / Pkg of 1
COMPARE
1
$57.50USD / Pkg of 1
COMPARE
1/16
$11.10USD / Pkg of 1
COMPARE
3/16
$11.10USD / Pkg of 1
COMPARE
3/8
$18.30USD / Pkg of 1
COMPARE
1/2
$23.50USD / Pkg of 1
COMPARE
3/4
$29.50USD / Pkg of 1
COMPARE
1
$42.75USD / Pkg of 1
Showing 1 - 20 of 29