This product is now discontinued, View Related Products
Comark HT100 Touch-Screen HACCP Data Logging Thermometer
Provides effective due diligence paperless records and tamper-proof data
  • Critical control points are monitored with programmable corrective actions
  • Store up to 65,000 readings in up to 100 separate folders
  • Download data to Microsoft® Excel® or export in PDF format
  • Accepts all type K thermocouple probes (order separately)
  • Water resistant with durable ABS plastic rubber boot
Comark’s HT100 touch-screen HACCP data logging thermometer is designed to collect, record, and store daily temperatures measurements within your receiving, storage, preparation, cooking, and holding areas. The meter can also be customized with user-defined checklists for temperature and nontemperature needs. Corrective actions can be logged to demonstrate your immediate response to any failures noted while doing your checklist.
The thermometer includes computer-based software that is easy and intuitive to use, allowing you to customize the program to be as simple or as detailed as you wish—record and set corrective actions for whatever is most important to you. The software can be downloaded from the Comark website at no charge.
Other features include 3” color touch screen, °F /°C selection, password protection, and low-battery indicator.
The HT100 HACCP data logging thermometer is also available with either a penetration probe with 1.5-mm tip or a faster responding probe with 1.1-mm microtip.
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
COMARK HT100 HACCP-Series Touch Data Logging Thermometer EW-90025-61 Mfr # HT100
COMPARE
999 Thermocouple
Call For Price
COMARK HtT100/PK15 HACCP-Series Touch Data Logging Thermometer with Pk15 Thin Penetration Probe EW-90025-62 Mfr # HT100/PK15
COMPARE
999 Thermocouple
Call For Price
COMARK HT100/PK19 HACCP-Series Touch Data Logging Thermometer with Pk19 Penetration Probe EW-90025-63 Mfr # HT100/PK19
COMPARE
999 Thermocouple